Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetamidothiazole 98.0+%, TCI America™
SDP

Catalog No. A147225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A147225G 25 g
A14725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A147225G Supplier TCI America Supplier No. A147225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2719-23-5
Molecular Formula C5H6N2OS
Molecular Weight (g/mol) 142.176
MDL Number MFCD00016892
InChI Key WXPLRSVMGRAIGW-UHFFFAOYSA-N
Synonym 2-acetamidothiazole, 2-acetylaminothiazole, n-thiazol-2-yl acetamide, acetamide, n-2-thiazolyl, n-1,3-thiazol-2-yl acetamide, n-2-thiazolylacetamide, thiazole, 2-acetamido, 2-acetaminothiazole, 2-acetamido-1,3-thiazole, n-thiazol-2-ylacetamide
PubChem CID 17625
IUPAC Name N-(1,3-thiazol-2-yl)acetamide
SMILES CC(=O)NC1=NC=CS1

Specifications

Melting Point 207°C
Color White-Yellow
Quantity 25 g
Formula Weight 142.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamidothiazole
EINECSNumber (5)-0207
RTECSNumber AC8800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.