Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2'-Acetoxyacetophenone, 98%

Click to view available options
Quantity:
5g
25g
100g
7250-94-4
C10H10O3
178.187
MFCD00017223
WWEDAIJJBDFOOK-UHFFFAOYSA-N
2'-acetoxyacetophenone,2-acetylphenyl acetate,ethanone, 1-2-acetyloxy phenyl,o-acetoxyacetophenone,2'-acetoxy acetophenone,maybridge1_002653,acmc-209on8,acetic acid 2-acetyl-phenyl ester,acetophenone, 2-hydroxy-, acetate 8ci
16716
(2-acetylphenyl) acetate
CC(=O)C1=CC=CC=C1OC(=O)C
7250-94-4
C10H10O3
178.187
MFCD00017223
WWEDAIJJBDFOOK-UHFFFAOYSA-N
2'-acetoxyacetophenone,2-acetylphenyl acetate,ethanone, 1-2-acetyloxy phenyl,o-acetoxyacetophenone,2'-acetoxy acetophenone,maybridge1_002653,acmc-209on8,acetic acid 2-acetyl-phenyl ester,acetophenone, 2-hydroxy-, acetate 8ci
16716
(2-acetylphenyl) acetate
CC(=O)C1=CC=CC=C1OC(=O)C
Melting Point 88°C to 91°C
Quantity 100g
Boiling Point 118°C (19mmHg)
Beilstein 1948938
Formula Weight 178.19
Percent Purity 98%
Chemical Name or Material 2'-Acetoxyacetophenone