Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Acetyl-1-methylpyrrole, 98%

Click to view available options
Quantity:
5g
25g
100g
932-16-1
C7H9NO
123.155
MFCD00003089
NZFLWVDXYUGFAV-UHFFFAOYSA-N
2-acetyl-1-methylpyrrole,n-methyl-2-acetylpyrrole,1-1-methyl-1h-pyrrol-2-yl ethanone,1-1-methylpyrrol-2-yl ethanone,1-methyl-2-acetylpyrrole,2-acetyl-n-methylpyrrole,ethanone, 1-1-methyl-1h-pyrrol-2-yl,methyl 1-methylpyrrol-2-yl ketone,1-1-methyl-1h-pyrrol-2-yl ethan-1-one,2-acetyl-1-methyl pyrrole
61240
CHEBI:59982
1-(1-methylpyrrol-2-yl)ethanone
CC(=O)C1=CC=CN1C
932-16-1
C7H9NO
123.155
MFCD00003089
NZFLWVDXYUGFAV-UHFFFAOYSA-N
2-acetyl-1-methylpyrrole,n-methyl-2-acetylpyrrole,1-1-methyl-1h-pyrrol-2-yl ethanone,1-1-methylpyrrol-2-yl ethanone,1-methyl-2-acetylpyrrole,2-acetyl-n-methylpyrrole,ethanone, 1-1-methyl-1h-pyrrol-2-yl,methyl 1-methylpyrrol-2-yl ketone,1-1-methyl-1h-pyrrol-2-yl ethan-1-one,2-acetyl-1-methyl pyrrole
61240
CHEBI:59982
1-(1-methylpyrrol-2-yl)ethanone
CC(=O)C1=CC=CN1C
Boiling Point 200°C to 202°C
Refractive Index 1.542
Density 1.04
Odor Pleasant
Quantity 25g
Flash Point 68°C (154°F)
Beilstein 111887
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 2-Acetyl-1-methylpyrrole

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 213-247-6

TSCA : Yes

Recommended Storage : Ambient temperatures