Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-5-chlorothiophene 99.0+%, TCI America™
SDP

Catalog No. A147125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A147125G 25 g
A14715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A147125G Supplier TCI America Supplier No. A147125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6310-09-4
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.62
MDL Number MFCD00005444
InChI Key HTZGPEHWQCRXGZ-UHFFFAOYSA-N
Synonym 2-acetyl-5-chlorothiophene, 1-5-chlorothiophen-2-yl ethanone, 1-5-chloro-2-thienyl ethanone, 5-chloro-2-acetylthiophene, ethanone, 1-5-chloro-2-thienyl, 5-chlor-2-acetyl thiophen, ketone, 5-chloro-2-thienyl methyl, 1-5-chlorothiophen-2-yl ethan-1-one, 5-chloro-2-thienyl methyl ketone, 2-chloro-5-acetylthiophene
PubChem CID 80572
IUPAC Name 1-(5-chlorothiophen-2-yl)ethanone
SMILES CC(=O)C1=CC=C(S1)Cl

Specifications

Melting Point 46°C
Boiling Point 118°C
Quantity 25 g
Formula Weight 160.62
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetyl-5-chlorothiophene
RTECSNumber OB1745000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.