Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-5-methylthiophene, 98%

Catalog No. AAA1128214
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 13679-74-8 | C7H8OS | 140.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13679-74-8
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.20
MDL Number MFCD00014529
InChI Key YOSDTJYMDAEEAZ-UHFFFAOYSA-N
Synonym 2-acetyl-5-methylthiophene, 1-5-methylthiophen-2-yl ethanone, ketone, methyl 5-methyl-2-thienyl, 1-5-methyl-2-thienyl ethan-1-one, 2-methyl-5-acetylthiophene, ethanone, 1-5-methyl-2-thienyl, methylthienylcetone, 1-5-methyl-2-thienyl ethanone, 5-methyl-2-acetylthiophene, methyl 5-methyl-2-thienyl ketone
PubChem CID 83655
IUPAC Name 1-(5-methylthiophen-2-yl)ethanone
SMILES CC(=O)C1=CC=C(C)S1

Specifications

Melting Point 24°C to 28°C
Density 1.119
Boiling Point 65°C to 67°C (1 mmHg)
Refractive Index 1.5625
Quantity 25 g
Beilstein 110854
Sensitivity Light sensitive
Formula Weight 140.2
Percent Purity 98%
Chemical Name or Material 2-Acetyl-5-methylthiophene
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 237-181-2
RTECSNumber OB4972000
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.