Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-6-methylpyridine, 98%

Catalog No. AAH65997MD
Click to view available options
Quantity:
250 mg
1 g
5 g

Catalog No. AAH65997MD

Supplier: Thermo Scientific Chemicals H65997MD

CAS: 6940-57-4 | C8H9NO | 135.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6940-57-4
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD06658293
InChI Key FPQMUQPPAYCAME-UHFFFAOYSA-N
Synonym 2-acetyl-6-methylpyridine, 1-6-methylpyridin-2-yl ethanone, 1-6-methylpyridin-2-yl ethan-1-one, ketone, methyl 6-methyl-2-pyridyl, 6-acetyl-2-picoline, 1-6-methyl-pyridin-2-yl-ethanone, 1-6-methyl-2-pyridinyl ethanone, 6-methyl-2-acetylpyridine, ethanone, 1-6-methyl-2-pyridinyl, 6-acetyl-2-methylpyridine
PubChem CID 138872
IUPAC Name 1-(6-methylpyridin-2-yl)ethanone
SMILES CC1=CC=CC(=N1)C(=O)C

Specifications

Color Yellow
Boiling Point 209°C
Flash Point 84°C (183°F)
Refractive Index 1.515
Quantity 250 mg
Beilstein 109564
Formula Weight 135.17
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Acetyl-6-methylpyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.