Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetylthiazole 98.0+%, TCI America™
SDP

Catalog No. A126525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A126525G 25 g
A12655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A126525G Supplier TCI America Supplier No. A126525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24295-03-2
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD00005324
InChI Key MOMFXATYAINJML-UHFFFAOYSA-N
Synonym 2-acetylthiazole, 1-1,3-thiazol-2-yl ethan-1-one, ethanone, 1-2-thiazolyl, 1-1,3-thiazol-2-yl ethanone, 2-acetyl thiazole, 1-2-thiazolyl ethanone, ketone, methyl 2-thiazolyl, 2-acetylthiazol, 2-acetyl-1,3-thiazole, 1-thiazol-2-yl-ethanone
PubChem CID 520108
IUPAC Name 1-(1,3-thiazol-2-yl)ethanone
SMILES CC(=O)C1=NC=CS1

Specifications

Color Yellow
Boiling Point 90°C
Quantity 25 g
Formula Weight 127.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Acetylthiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.