Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acrylamido-2-methylpropanesulfonic acid, 98%

Catalog No. AAA189240B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA189240B 1000 g
AAA1892430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA189240B Supplier Thermo Scientific Chemicals Supplier No. A189240B
Add to Cart

CAS: 15214-89-8 | C7H13NO4S | 207.24 g/mol

2-Acrylamide-2-methylpropanesulfonic acid has good complexion, adsorption, biological activity, surface activity, hydrolysis stability and thermal stability. It can be used in oil chemical, water treatment, synthetic fiber, printing and dyeing, plastics, water absorbing coatings, paper, bio-medical, magnetic materials and cosmetics industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acrylamide-2-methylpropanesulfonic acid has good complexion, adsorption, biological activity, surface activity, hydrolysis stability and thermal stability. It can be used in oil chemical, water treatment, synthetic fiber, printing and dyeing, plastics, water absorbing coatings, paper, bio-medical, magnetic materials and cosmetics industries.

Solubility
Highly soluble in water (1×106 mg/L at 25°C ).

Notes
Hygroscopic. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15214-89-8
Molecular Formula C7H13NO4S
Molecular Weight (g/mol) 207.24
MDL Number MFCD00007522
InChI Key XHZPRMZZQOIPDS-UHFFFAOYSA-N
Synonym 2-acrylamido-2-methylpropanesulfonic acid, 2-acrylamido-2-methyl-1-propanesulfonic acid, 2-acrylamide-2-methylpropanesulfonic acid, 1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino, polyacrylamidomethylpropane sulfonic acid, unii-490hqe5ki5, 2-acrylamido-2-methylpropanesulfonate, 2-acrylamido-2-methylpropanesulphonic acid, 2-acrylamido-2-methylpropane sulfonic acid, 2-acrylamido-2-methylpropane-1-sulfonic acid
PubChem CID 65360
IUPAC Name 2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid
SMILES CC(C)(CS(O)(=O)=O)NC(=O)C=C

Specifications

Melting Point ∼195°C (decomposition)
Density 1.1
Quantity 1000 g
UN Number UN2585
Beilstein 1946464
Sensitivity Hygroscopic
Solubility Information Highly soluble in water (1×106mg/L at 25°C ).
Formula Weight 207.25
Percent Purity 98%
Chemical Name or Material 2-Acrylamido-2-methylpropanesulfonic acid
Hazard Category H302-H312-H318
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYL SULFONIC ACIDS, SOLID
EINECSNumber 239-268-0
RTECSNumber TZ6658000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.