Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Adamantanone 98.0+%, TCI America™
SDP

Catalog No. A071925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A071925G 25 g
A07195G 5 g
A0719100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A071925G Supplier TCI America Supplier No. A071925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 700-58-3
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00074737
InChI Key IYKFYARMMIESOX-UHFFFAOYSA-N
Synonym 2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one
PubChem CID 64151
ChEBI CHEBI:40611
IUPAC Name adamantan-2-one
SMILES C1C2CC3CC1CC(C2)C3=O

Specifications

Color White
Quantity 25 g
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Adamantanone
EINECSNumber (4)-1894
RTECSNumber AU5018000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.