Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Allyl-L-glycine, 95%, Thermo Scientific™

Catalog No. AAH5209703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5209703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5209703 Supplier Thermo Scientific Chemicals Supplier No. H5209703

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 195316-72-4
Molecular Formula C5H10ClNO2
Molecular Weight (g/mol) 151.59
MDL Number MFCD00002627
InChI Key DIDZZOWASZMNQW-WCCKRBBISA-N
Synonym s-2-aminopent-4-enoic acid hydrochloride, l-2-allylglycine hydrochloride, s---2-amino-4-pentenoic acid hydrochloride, 2s-2-aminopent-4-enoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, 2s, l-allylglycine hydrochloride, l-alpha-allylglycine hydrochloride, s-2-amino-4-pentenoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, s
PubChem CID 44630067
IUPAC Name (2S)-2-aminopent-4-enoic acid;hydrochloride
SMILES C=CCC(C(=O)O)N.Cl

Specifications

Quantity 1 g
Formula Weight 115.13
Percent Purity 95%
Chemical Name or Material 2-Allyl-L-glycine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.