Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-2-Amino-1-butanol, 97%

Catalog No. p-7027557
Click to view available options
Quantity:
100 mL
500 mL

CAS: 96-20-8 | C4H11NO | 89.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-20-8
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00008095
InChI Key JCBPETKZIGVZRE-UHFFFAOYSA-N
Synonym 2-amino-1-butanol, 2-aminobutanol, dl-2-amino-1-butanol, 1-butanol, 2-amino, 2-aminobutyl alcohol, 1-hydroxy-2-butylamine, butanol-2-amine, 1-hydroxymethyl propylamine, 2-amino-1-hydroxybutane, 2-amino-n-butyl alcohol
PubChem CID 22129
IUPAC Name 2-aminobutan-1-ol
SMILES CCC(CO)N

Specifications

Melting Point -2°C
Density 0.944
pH 11.1
Boiling Point 176°C to 178°C
Flash Point 89°C (192°F)
Odor Amine-like
Refractive Index 1.4525
Quantity 100 mL
UN Number UN2735
Beilstein 1098274
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,427
Formula Weight 89.14
Percent Purity 97%
Chemical Name or Material (+/-)-2-Amino-1-butanol
Show More Show Less
Hazard Category H227-H290-H302-H312-H314-H332-H335
Hazard Statement GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P233-P234-P240-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P374-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 202-488-2
RTECSNumber EK9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.