Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-2-thiazoline, 97%

Catalog No. p-4228344
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle

CAS: 1779-81-3 | C3H6N2S | 102.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1779-81-3
Molecular Formula C3H6N2S
Molecular Weight (g/mol) 102.15
InChI Key REGFWZVTTFGQOJ-UHFFFAOYSA-N
Synonym 2-aminothiazoline, 2-amino-2-thiazoline, 2-iminothiazolidine, 4,5-dihydrothiazol-2-amine, 2-thiazolidinimine, 2-amino-4,5-dihydrothiazole, 2-thiazolamine, 4,5-dihydro, 2-thiazoline, 2-amino, usaf pd-57, 2-amino-4,5-dihydrothiazoline
PubChem CID 15689
ChEBI CHEBI:40889
IUPAC Name 4,5-dihydro-1,3-thiazol-2-amine
SMILES C1CSC(=N1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 77°C to 81°C
Color White
Boiling Point 221.8°C
Flash Point 87.9°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 102.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-2-thiazoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-224-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.