Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3,5-dibromobenzonitrile, 97%, Thermo Scientific™

Catalog No. AAH6415503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6415503 1 g
AAH6415506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6415503 Supplier Thermo Scientific Chemicals Supplier No. H6415503

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68385-95-5
Molecular Formula C7H4Br2N2
Molecular Weight (g/mol) 275.93
MDL Number MFCD00179571
InChI Key WLZCMGXAEXFAQA-UHFFFAOYSA-N
Synonym 3,5-dibromo-2-aminobenzonitrile, 2-amino-3,5-dibromo-benzonitrile, benzonitrile,2-amino-3,5-dibromo, buttpark 37\12-53, 3,5-dibromoanthranilonitrile, 2-amino-3,5-dibromobenzenecarbonitrile, pubchem14450, 2-amino-3,5-dibromobenzonitrile
PubChem CID 2734082
IUPAC Name 2-amino-3,5-dibromobenzonitrile
SMILES NC1=C(Br)C=C(Br)C=C1C#N

Specifications

Melting Point 152°C to 156°C (literature)
Quantity 1 g
Beilstein 2834670
Formula Weight 275.93
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-3,5-dibromobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.