Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3,5-dichloropyridine, 97%

Catalog No. p-54791
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159380050 5 g Glass Bottle
AC159380250 25 g Plastic Bottle
AC159381000 100 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC159380050 Supplier Thermo Scientific Chemicals Supplier No. 159380050
Only null left
Add to Cart

CAS: 4214-74-8 | C5H4Cl2N2 | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4214-74-8
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.00
MDL Number MFCD00006313
InChI Key OCWBGKZFOYMCCN-UHFFFAOYSA-N
Synonym 2-amino-3,5-dichloropyridine, 3,5-dichloro-2-pyridinamine, 2-pyridinamine, 3,5-dichloro, 3,5-dichloro-3-pyridylamine, 3,5-dichloro-2-aminopyridine, 3,5-dichloro-2-pyridylamine, 3,5-dichloropyridin-2-ylamine, pyridine, 2-amino-3,5-dichloro, pubchem13504, acmc-209jnv
PubChem CID 77886
IUPAC Name 3,5-dichloropyridin-2-amine
SMILES NC1=NC=C(Cl)C=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 79°C to 83°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula Cl2(C5H2N)NH2
Quantity 5 g
Beilstein 22, 430
Formula Weight 163.01
Percent Purity ≥96.0% (HPLC)
Physical Form Crystals or Powder
Chemical Name or Material 2-Amino-3, 5-dichloropyridine, 97%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if inhaled. Causes skin irritation. May cause respiratory irritation. Harmful if swallowed. Causes serious eye irritation. Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 224-143-5
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.