Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-benzyloxypyridine 98.0+%, TCI America™
SDP

Catalog No. A208025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A208025G 25 g
A20805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A208025G Supplier TCI America Supplier No. A208025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24016-03-3
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.24
MDL Number MFCD00006316
InChI Key NMCBWICNRJLKKM-UHFFFAOYSA-N
Synonym 2-amino-3-benzyloxypyridine, 3-benzyloxy pyridin-2-amine, 3-benzyloxy-2-pyridylamine, 2-pyridinamine, 3-phenylmethoxy, 2-amino-3-benzyloxy pyridine, 3ip, 3-phenylmethoxy-2-pyridylamine, 3-benzyloxy pyridin-2-amin, 3fty
PubChem CID 90334
IUPAC Name 3-(benzyloxy)pyridin-2-amine
SMILES NC1=NC=CC=C1OCC1=CC=CC=C1

Specifications

Melting Point 97°C
Color Yellow
Quantity 25 g
Formula Weight 200.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-benzyloxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.