Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-bromo-5-methylpyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
17282-00-7
C6H7BrN2
187.04
MFCD00068231
NDPKXEWDWTZBDG-UHFFFAOYSA-N
2-amino-3-bromo-5-methylpyridine,2-amino-3-bromo-5-picoline,6-amino-5-bromo-3-picoline,3-bromo-5-methyl-2-pyridinamine,2-amino-5-methyl-3-bromopyridine,2-pyridinamine, 3-bromo-5-methyl,3-bromo-5-methyl-pyridin-2-ylamine,3-bromo-5-methyl-2-pyridylamine,3-bromo-5-methylpyridin-2-ylamine,3-bromo-5-methylpyridin-1-ium-2-amine
519380
3-bromo-5-methylpyridin-2-amine
CC1=CC(=C(N=C1)N)Br
17282-00-7
C6H7BrN2
187.04
MFCD00068231
NDPKXEWDWTZBDG-UHFFFAOYSA-N
2-amino-3-bromo-5-methylpyridine,2-amino-3-bromo-5-picoline,6-amino-5-bromo-3-picoline,3-bromo-5-methyl-2-pyridinamine,2-amino-5-methyl-3-bromopyridine,2-pyridinamine, 3-bromo-5-methyl,3-bromo-5-methyl-pyridin-2-ylamine,3-bromo-5-methyl-2-pyridylamine,3-bromo-5-methylpyridin-2-ylamine,3-bromo-5-methylpyridin-1-ium-2-amine
519380
3-bromo-5-methylpyridin-2-amine
CC1=CC(=C(N=C1)N)Br
Color Beige to Yellow or Brown
Melting Point 55.0°C to 57.0°C
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,chloroform
Formula Weight 187.04
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Amino-3-bromo-5-methylpyridine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning