Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-methylbenzoic acid, 98%

Catalog No. AAB2423006
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAB2423006

Supplier: Thermo Scientific Chemicals B2423006

Only null left
Add to Cart

CAS: 4389-45-1 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4389-45-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007745
InChI Key WNAJXPYVTFYEST-UHFFFAOYSA-N
Synonym 3-methylanthranilic acid, 2-amino-3-methyl-benzoic acid, 2-amino-3-methylbenzoicacid, 3-methyl-2-aminobenzoic acid, benzoic acid, 2-amino-3-methyl, 2-amino-m-toluic acid, 2-amino-3-methyl benzoic acid, m-toluic acid, 2-amino, unii-8rgx6sqe9n, 2-amino-3-methylbenzoate
PubChem CID 78101
ChEBI CHEBI:80574
IUPAC Name 2-amino-3-methylbenzoic acid
SMILES CC1=CC=CC(=C1N)C(=O)O

Specifications

Melting Point 174°C to 177°C
Quantity 5 g
Beilstein 2359694
Formula Weight 151.16
Percent Purity 98%
Chemical Name or Material 2-Amino-3-methylbenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 224-505-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.