Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-methylpyrazine, 95%, Thermo Scientific™

Catalog No. p-7041187
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19838-08-5
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.132
MDL Number MFCD08705770
InChI Key VQPHZDDLWFHRHR-UHFFFAOYSA-N
Synonym 2-amino-3-methylpyrazine, 3-methyl-2-pyrazinamine, pyrazinamine, 3-methyl, 2-pyrazinamine, 3-methyl, 3-methyl-2-aminopyrazine, pubchem19617, acmc-1cacy, 2-pyrazinamine,3-methyl, 3-methylpyrazin-2-ylamine, 2-amino-3-methyl-pyrazine
PubChem CID 302284
IUPAC Name 3-methylpyrazin-2-amine
SMILES CC1=NC=CN=C1N

Specifications

Color Yellow
Quantity 1 g
Formula Weight 109.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-3-methylpyrazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.