Alfa Aesar™ 2-Amino-3-methylpyridine, 97%

Manufacturer: Alfa Aesar™ A1022422

Quantity 100g

 View more versions of this product

Catalog No. AAA1022422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1603-40-3
    Molecular Formula C6H8N2
    Formula Weight 108.14
    MDL Number MFCD00006320
    UN Number UN2811
    Beilstein 107892
    Synonym 2-Amino-3-picoline; 3-Methyl-2-pyridinamine
    Chemical Name or Material 2-Amino-3-methylpyridine

    Specifications

    Formula Weight 108.14
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P307+P311
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed: Call a POISON CENTER or doctor/physician.
    Health Hazard 2 GHS H Statement
    H301-H310-H315-H319-H335
    Toxic if swallowed.
    Fatal in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Boiling Point 221° to 222°C
    Quantity 100g
    Beilstein 107892
    TSCA TSCA
    Molecular Formula C6H8N2
    UN Number UN2811
    Flash Point 118°C (244°F)
    Melting Point 29° to 32°C
    MDL Number MFCD00006320
    Synonym 2-Amino-3-picoline; 3-Methyl-2-pyridinamine
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number US1850000
    CAS 1603-40-3
    Odor Unpleasant
    Health Hazard 1 Danger
    Chemical Name or Material 2-Amino-3-methylpyridine
    Assay 97%

    GHS H Statement
    H301-H310-H315-H319-H335
    Toxic if swallowed.
    Fatal in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P307+P311
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed: Call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit