Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-methylpyridine, 97%

Catalog No. p-7080622
Click to view available options
Quantity:
25 g
100 g

CAS: 1603-40-3 | C6H8N2 | 108.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1603-40-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006320
InChI Key RGDQRXPEZUNWHX-UHFFFAOYSA-N
Synonym 2-amino-3-picoline, 2-amino-3-methylpyridine, 2-pyridinamine, 3-methyl, 3-methyl-2-pyridinamine, 3-methyl-2-aminopyridine, 3-methyl-2-pyridylamine, 3-picoline, 2-amino, 2-amino-3-methyl pyridine, 2-amino-beta-picoline, pyridine, 2-amino-3-methyl
PubChem CID 15347
IUPAC Name 3-methylpyridin-2-amine
SMILES CC1=C(N=CC=C1)N

Specifications

Melting Point 29°C to 32°C
Density 1.073
Boiling Point 221°C to 222°C
Flash Point 118°C (244°F)
Odor Unpleasant
Refractive Index 1.5825
Quantity 25 g
UN Number UN2811
Beilstein 107892
Sensitivity Hygroscopic
Formula Weight 108.14
Percent Purity 97%
Chemical Name or Material 2-Amino-3-methylpyridine
Show More Show Less
Hazard Category H301+H311-H315-H319-H335
Hazard Statement GHS H Statement
H301-H310-H315-H319-H335
Toxic if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 216-501-4
RTECSNumber US1850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.