Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-nitrophenol, 98%

Catalog No. p-3211588
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 603-85-0 | C6H6N2O3 | 154.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-85-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00010875
InChI Key KUCWUAFNGCMZDB-UHFFFAOYSA-N
Synonym phenol, 2-amino-3-nitro, 2-hydroxy-6-nitroaniline, ccris 2544, unii-a7f97zpu2w, 3-nitro-2-aminophenol, 2-amino-3-nitro-phenol, a7f97zpu2w, 2-amino-3-hydroxynitrobenzene, 1-hydroxy-2-amino-3-nitrobenzene, 2-amino-nitrophenol
PubChem CID 4115495
IUPAC Name 2-amino-3-nitrophenol
SMILES NC1=C(O)C=CC=C1[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 215.0°C
Color Red to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC6H3(NO2)OH
Quantity 5 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 154.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-3-nitrophenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective g
EINECSNumber 210-060-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.