Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-(trifluoromethyl)pyridine-5-boronic acid pinacol ester, 96%

Catalog No. p-7036490
Click to view available options
Quantity:
250 mg
1 g

CAS: 947249-01-6 | C12H16BF3N2O2 | 288.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947249-01-6
Molecular Formula C12H16BF3N2O2
Molecular Weight (g/mol) 288.08
MDL Number MFCD12923414
InChI Key OQZKROYNCVLJKM-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-trifluoromethyl pyridin-2-amine, 2-amino-3-trifluoromethyl pyridine-5-boronic acid pinacol ester, 6-amino-5-trifluoromethyl pyridine-3-boronic acid pinacol ester, 6-amino-5-trifluoromethyl pyridin-3-yl boronic acid pinacol ester, 3-trifluoromethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 5-4,4,5,5-tetra methyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl-pyridin-2-ylamine, 5-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl-3-trifluoromethyl-2-pyridylamine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl-pyridin-2-ylamine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl pyridin-2-amine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl-pyridin-2-yl-amine
PubChem CID 46864103
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridin-2-amine
SMILES CC1(C)OB(OC1(C)C)C1=CC(=C(N)N=C1)C(F)(F)F

Specifications

Melting Point 132°C to 134°C
Quantity 250 mg
Formula Weight 288.07
Percent Purity 96%
Chemical Name or Material 2-Amino-3-(trifluoromethyl)pyridine-5-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.