Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-chloro-6-methoxypyrimidine, 99%, Thermo Scientific™

Catalog No. AAH6670322
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6670322 100 g
AAH6670336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6670322 Supplier Thermo Scientific Chemicals Supplier No. H6670322

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5734-64-5
Molecular Formula C5H6ClN3O
Molecular Weight (g/mol) 159.57
MDL Number MFCD00233535
InChI Key VFEYBTFCBZMBAU-UHFFFAOYSA-N
Synonym 2-amino-4-chloro-6-methoxypyrimidine, 2-pyrimidinamine, 4-chloro-6-methoxy, 2-amino-6-chloro-4-methoxypyrimidine, pyrimidine, 2-amino-4-chloro-6-methoxy, 2-amino-4-methoxy-6-chloro pyrimidine, 4-chloro-6-methoxy-2-pyrimidinamine, 2-amino-4-chloro-6-methoxylpyrimidine, 2-amino-4-chloro-6-methoxy pyrimidine, 2-amino-4-chloro-6-methoxy-pyrimidine, 4-chloro-6-methoxy-pyrimidin-2-ylamine
PubChem CID 79811
IUPAC Name 4-chloro-6-methoxypyrimidin-2-amine
SMILES COC1=CC(Cl)=NC(N)=N1

Specifications

Melting Point 168°C to 173°C
Quantity 100 g
Beilstein 127277
Formula Weight 159.57
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 2-Amino-4-chloro-6-methoxypyrimidine
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 401-050-9
RTECSNumber UV6260335
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.