Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-chloro-6-methoxypyrimidine 95.0+%, TCI America™
SDP

Catalog No. A152025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A152025G 25 g
A15205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A152025G Supplier TCI America Supplier No. A152025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5734-64-5
Molecular Formula C5H6ClN3O
Molecular Weight (g/mol) 159.57
MDL Number MFCD00233535
InChI Key VFEYBTFCBZMBAU-UHFFFAOYSA-N
Synonym 2-amino-4-chloro-6-methoxypyrimidine, 2-pyrimidinamine, 4-chloro-6-methoxy, 2-amino-6-chloro-4-methoxypyrimidine, pyrimidine, 2-amino-4-chloro-6-methoxy, 2-amino-4-methoxy-6-chloro pyrimidine, 4-chloro-6-methoxy-2-pyrimidinamine, 2-amino-4-chloro-6-methoxylpyrimidine, 2-amino-4-chloro-6-methoxy pyrimidine, 2-amino-4-chloro-6-methoxy-pyrimidine, 4-chloro-6-methoxy-pyrimidin-2-ylamine
PubChem CID 79811
IUPAC Name 4-chloro-6-methoxypyrimidin-2-amine
SMILES COC1=CC(Cl)=NC(N)=N1

Specifications

Melting Point 170°C
Color Yellow
Quantity 25 g
Formula Weight 159.57
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chloro-6-methoxypyrimidine
RTECSNumber UV6260335
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.