Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-chlorothiophenol, 96%

Manufacturer:  Alfa Aesar™ B2564014

Quantity 25g

 View more versions of this product

Catalog No. AAB2564014


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1004-00-8
159.64
UN1759
2-Amino-4-chlorothiophenol
C6H6ClNS
MFCD00792528
2-Amino-4-chlorobenzenethiol

Specifications

1004-00-8
25g
C6H6ClNS
MFCD00792528
2-Amino-4-chlorobenzenethiol
96%
52° to 59°C
Unpleasant
159.64
UN1759
2-Amino-4-chlorothiophenol

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit