Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-hydroxy-6-methylpyrimidine 98.0+%, TCI America™
SDP

Catalog No. A1204250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1204250G 250 g
A120425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1204250G Supplier TCI America Supplier No. A1204250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3977-29-5
Molecular Formula C5H7N3O
Molecular Weight (g/mol) 125.131
MDL Number MFCD00006095
InChI Key KWXIPEYKZKIAKR-UHFFFAOYSA-N
Synonym 2-amino-4-hydroxy-6-methylpyrimidine, 2-amino-6-methylpyrimidin-4-ol, 6-methylisocytosine, mecytosine, 2-amino-6-methylpyrimidin-4 3h-one, superacil, superacyl, 4-methylisocytosine, 2-amino-6-hydroxy-4-methylpyrimidine, 2-amino-6-methyl-4-pyrimidinol
PubChem CID 1532
ChEBI CHEBI:58959
IUPAC Name 2-amino-6-methyl-1H-pyrimidin-4-one
SMILES CC1=CC(=O)N=C(N1)N

Specifications

Color White-Yellow
Quantity 250 g
Formula Weight 125.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-hydroxy-6-methylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.