Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-hydroxybenzothiazole 98.0+%, TCI America™
SDP

Catalog No. A30011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30011G 1 g
A30015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30011G Supplier TCI America Supplier No. A30011G

Chemical Identifiers

CAS 7471-03-6
Molecular Formula C7H6N2OS
Molecular Weight (g/mol) 166.20
MDL Number MFCD00160060
InChI Key PFQJPSASUCHKRO-UHFFFAOYSA-N
Synonym 2-Aminobenzo[d]thiazol-4-ol
PubChem CID 24057
IUPAC Name 2-amino-1,3-benzothiazol-4-ol
SMILES C1=CC(=C2C(=C1)SC(=N2)N)O

Specifications

Melting Point 184°C
Quantity 1 g
Formula Weight 166.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-hydroxybenzothiazole
RTECSNumber DL1800000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.