Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-methylbenzothiazole, 98%

Catalog No. p-7081030
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1477-42-5 | C8H8N2S | 164.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1477-42-5
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00005793
InChI Key GRIATXVEXOFBGO-UHFFFAOYSA-N
Synonym 2-amino-4-methylbenzothiazole, 4-methylbenzo d thiazol-2-amine, 2-benzothiazolamine, 4-methyl, 4-methyl-2-aminobenzothiazole, 4-methyl-2-benzothiazolamine, benzothiazole, 2-amino-4-methyl, 4-methylbenzothiazol-2-ylamine, 2-amino-4-methyl benzothiazole, 4-methyl-2-aminobenzothiozole, unii-45v5f1b9z8
PubChem CID 15132
IUPAC Name 4-methyl-1,3-benzothiazol-2-amine
SMILES CC1=C2N=C(N)SC2=CC=C1

Specifications

Melting Point 137°C to 139°C
Quantity 5 g
Beilstein 129346
Sensitivity Moisture sensitive
Formula Weight 164.22
Percent Purity 98%
Chemical Name or Material 2-Amino-4-methylbenzothiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 216-028-3
RTECSNumber DL2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.