Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-methylpyrimidine, 97%

Catalog No. AAA1608118
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1608118 50 g
AAA1608109 10 g
AAA1608130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1608118 Supplier Thermo Scientific Chemicals Supplier No. A1608118
Add to Cart

CAS: 108-52-1 | C5H7N3 | 109.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-52-1
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.132
MDL Number MFCD00006101
InChI Key GHCFWKFREBNSPC-UHFFFAOYSA-N
Synonym 2-amino-4-methylpyrimidine, 2-pyrimidinamine, 4-methyl, 6-methyl-2-pyrimidinamine, pyrimidine, 2-amino-4-methyl, 4-methyl-2-pyrimidinamine, 2-amino-4-methyl-pyrimidine, 4-methylpyrimidin-2-ylamine, 4-methylpyrimidine-2-ylamine, 4-methyl-pyrimidin-2-ylamine, 2-amino-6-methylpyrimidine
PubChem CID 7939
IUPAC Name 4-methylpyrimidin-2-amine
SMILES CC1=NC(=NC=C1)N

Specifications

Melting Point 158°C to 161°C
Quantity 50 g
Beilstein 108506
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2-Amino-4-methylpyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-591-5
RTECSNumber UV6485000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.