Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-amino-4-picoline, 98%

Catalog No. p-3735517
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

CAS: 695-34-1 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 695-34-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006321
InChI Key ORLGLBZRQYOWNA-UHFFFAOYSA-N
Synonym 2-amino-4-methylpyridine, 2-amino-4-picoline, 2-pyridinamine, 4-methyl, ascensil, 4-methyl-2-pyridinamine, 4-methyl-2-pyridylamine, 4-methyl-2-aminopyridine, 4-picoline, 2-amino, w 45 raschig, aminton
PubChem CID 1533
IUPAC Name 4-methylpyridin-2-amine
SMILES CC1=CC(=NC=C1)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 96°C to 101°C
Color Beige to Yellow or Brown
Boiling Point 230°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 22, V,9, 325
Merck Index 15, 462
Solubility Information Solubility in water: 41g/L (20°C). Other solubilities: soluble in lower alcohols,slightly soluble in petroleum ether
Formula Weight 108.14
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material 2-Amino-4-picoline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Toxic in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-780-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.