Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-Amino-4-tert-butylphenol, 97%
Click to view available options
Quantity:
5g
25g
Specifications
Quantity | 25g |
Melting Point | 162°C to 164°C |
Formula Weight | 165.23 |
Percent Purity | 97% |
Chemical Name or Material | 2-Amino-4-tert-butylphenol |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 214-844-1
TSCA : No
Recommended Storage : Ambient temperatures