Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-thiazoleacetic acid, 97%

Click to view available options
Quantity:
25g
100g
29676-71-9
C5H6N2O2S
158.175
MFCD00010068
DYCLHZPOADTVKK-UHFFFAOYSA-N
2-amino-4-thiazoleacetic acid,2-2-aminothiazol-4-yl acetic acid,2-aminothiazol-4-acetic acid,2-aminothiazole-4-yl acetic acid,4-thiazoleacetic acid, 2-amino,2-amino-1,3-thiazol-4-yl acetic acid,2-aminothiazole-4-yl-acetic acid,2-aminothiazole-4-acetic acid,unii-qsh3o4tnjt,2-2-amino-1,3-thiazol-4-yl acetic acid
34665
2-(2-amino-1,3-thiazol-4-yl)acetic acid
C1=C(N=C(S1)N)CC(=O)O
29676-71-9
C5H6N2O2S
158.175
MFCD00010068
DYCLHZPOADTVKK-UHFFFAOYSA-N
2-amino-4-thiazoleacetic acid,2-2-aminothiazol-4-yl acetic acid,2-aminothiazol-4-acetic acid,2-aminothiazole-4-yl acetic acid,4-thiazoleacetic acid, 2-amino,2-amino-1,3-thiazol-4-yl acetic acid,2-aminothiazole-4-yl-acetic acid,2-aminothiazole-4-acetic acid,unii-qsh3o4tnjt,2-2-amino-1,3-thiazol-4-yl acetic acid
34665
2-(2-amino-1,3-thiazol-4-yl)acetic acid
C1=C(N=C(S1)N)CC(=O)O
Quantity 25g
Melting Point ~130°C (decomposition)
Beilstein 127415
Formula Weight 158.18
Percent Purity 97%
Chemical Name or Material 2-Amino-4-thiazoleacetic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning