Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-5-bromo-2'-fluorobenzophenone, 95%

Click to view available options
Quantity:
1g
5g
1479-58-9
C13H9BrFNO
294.123
MFCD00038380
XCOKDXNGCQXFCV-UHFFFAOYSA-N
2-amino-5-bromo-2'-fluorobenzophenone,2-amino-2'-fluoro-5-bromobenzophenone,2-amino-5-bromophenyl 2-fluorophenyl methanone,methanone, 2-amino-5-bromophenyl 2-fluorophenyl,4-bromo-2-2-fluorobenzoyl aniline,2-amino-5-bromophenyl 2-fluorophenyl ketone,2-amino-5-bromo-2-fluorobenzophenone,2-amino-5-bromo-phenyl-2-fluoro-phenyl-methanone,2-amino-2-fluoro-5-bromobenzophenone,2-amino-5-bromo-2'-fluoro benzophenone
73865
(2-amino-5-bromophenyl)-(2-fluorophenyl)methanone
C1=CC=C(C(=C1)C(=O)C2=C(C=CC(=C2)Br)N)F
1479-58-9
C13H9BrFNO
294.123
MFCD00038380
XCOKDXNGCQXFCV-UHFFFAOYSA-N
2-amino-5-bromo-2'-fluorobenzophenone,2-amino-2'-fluoro-5-bromobenzophenone,2-amino-5-bromophenyl 2-fluorophenyl methanone,methanone, 2-amino-5-bromophenyl 2-fluorophenyl,4-bromo-2-2-fluorobenzoyl aniline,2-amino-5-bromophenyl 2-fluorophenyl ketone,2-amino-5-bromo-2-fluorobenzophenone,2-amino-5-bromo-phenyl-2-fluoro-phenyl-methanone,2-amino-2-fluoro-5-bromobenzophenone,2-amino-5-bromo-2'-fluoro benzophenone
73865
(2-amino-5-bromophenyl)-(2-fluorophenyl)methanone
C1=CC=C(C(=C1)C(=O)C2=C(C=CC(=C2)Br)N)F
Melting Point 100°C to 104°C
Quantity 5g
Formula Weight 294.12
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Amino-5-bromo-2'-fluorobenzophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 216-037-2

TSCA : No

Recommended Storage : Ambient temperatures