Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-bromobenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. A30631G
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 29124-57-0
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD10696879
InChI Key VBYZWJMZASVGNB-UHFFFAOYSA-N
Synonym 2-amino-5-bromobenzenecarbaldehyde, 5-bromoanthranilaldehyde, 4-bromo-2-formylaniline, benzaldehyde,2-amino-5-bromo, benzaldehyde, 2-amino-5-bromo, pubchem19693, aminobromobenzenecarbaldehyde, 2-amino-5-bromobenzaidehyde, 2-amino-5-bromo-benzaldehyde
PubChem CID 23510475
IUPAC Name 2-amino-5-bromobenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)N

Specifications

Melting Point 78°C
Color Yellow
Quantity 1 g
Formula Weight 200.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromobenzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.