Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-bromobenzothiazole, 97%

Catalog No. p-7052679
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 20358-03-6 | C7H5BrN2S | 229.095 g/mol

2-Amino-5-bromobenzothiazole is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-bromobenzothiazole is used as pharmaceutical intermediates

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20358-03-6
Molecular Formula C7H5BrN2S
Molecular Weight (g/mol) 229.095
MDL Number MFCD00577806
InChI Key ZPUJTWBWSOOMRP-UHFFFAOYSA-N
Synonym 2-amino-5-bromobenzothiazole, 5-bromobenzo d thiazol-2-amine, 5-bromo-benzothiazol-2-ylamine, 2-benzothiazolamine, 5-bromo, 5-bromo-2-benzothiazolamine, 2-amino-5-bromo-1,3-benzothiazole, pubchem21702, acmc-209f9a, 2-amino-5-bromo benzothiazole, 5-bromobenzothiazole-2-ylamine
PubChem CID 13775785
IUPAC Name 5-bromo-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1Br)N=C(S2)N

Specifications

Melting Point 194°C to 198°C
Color White to Brown
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 229.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-5-bromobenzothiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.