Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-bromopyrazine, 97%

Catalog No. p-7029942
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2746503 1 g
AAH2746506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2746503 Supplier Thermo Scientific Chemicals Supplier No. H2746503
Only null left
Add to Cart

CAS: 59489-71-3 | C4H4BrN3 | 174.001 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59489-71-3
Molecular Formula C4H4BrN3
Molecular Weight (g/mol) 174.001
MDL Number MFCD00235015
InChI Key KRRTXVSBTPCDOS-UHFFFAOYSA-N
Synonym 2-amino-5-bromopyrazine, 5-bromo-2-pyrazinamine, 5-bromo-pyrazin-2-ylamine, 2-bromo-5-aminopyrazine, pyrazinamine, 5-bromo, 5-amino-2-bromopyrazine, 5-bromo-2-aminopyrazine, 5-bromopyrazin-2-ylamine, 5-bromopyrazinamine, 2-amino-5-bromo-pyrazine
PubChem CID 599539
IUPAC Name 5-bromopyrazin-2-amine
SMILES C1=C(N=CC(=N1)Br)N

Specifications

Melting Point 113°C to 117°C
Quantity 1 g
Formula Weight 174
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromopyrazine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.