Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-fluorobenzothiazole, 98%

Catalog No. p-7033844
Click to view available options
Quantity:
250 mg
1 g

CAS: 20358-07-0 | C7H5FN2S | 168.189 g/mol

2-Amino-5-fluorobenzothiazole is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-fluorobenzothiazole is used as organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20358-07-0
Molecular Formula C7H5FN2S
Molecular Weight (g/mol) 168.189
MDL Number MFCD05664560
InChI Key YHBIGBYIUMCLJS-UHFFFAOYSA-N
PubChem CID 821203
IUPAC Name 5-fluoro-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1F)N=C(S2)N

Specifications

Quantity 250 mg
Synonym 2-amino-5-fluorobenzothiazole, 5-fluorobenzo d thiazol-2-amine, 2-benzothiazolamine, 5-fluoro, 5-fluoro-2-benzothiazolamine, 2-benzothiazolamine,5-fluoro, 5-fluorobenzothiazol-2-ylamine, pubchem21850, acmc-20a67y, 2-amino-5-fluorobenzothiozole, 5-fluoro-2-aminobenzothiazole
Solubility Information Soluble in methanol.
Formula Weight 168.19
Percent Purity 98%
Chemical Name or Material 2-Amino-5-fluorobenzothiazole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.