Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-iodobenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
5326-47-6
C7H6INO2
263.03
MFCD00007849
GOLGILSVWFKZRQ-UHFFFAOYSA-N
5-iodoanthranilic acid,benzoic acid, 2-amino-5-iodo,5-iodoanthranil acid,anthranilic acid, 5-iodo,2-amino-5-iodo-benzoic acid,pubchem2543,5-iodo anthranilic acid,acmc-1ays8,2-amino-5-iodobenzoicacid,5-iodo-2-aminobenzoic acid
72911
2-amino-5-iodobenzoic acid
C1=CC(=C(C=C1I)C(=O)O)N
5326-47-6
C7H6INO2
263.03
MFCD00007849
GOLGILSVWFKZRQ-UHFFFAOYSA-N
5-iodoanthranilic acid,benzoic acid, 2-amino-5-iodo,5-iodoanthranil acid,anthranilic acid, 5-iodo,2-amino-5-iodo-benzoic acid,pubchem2543,5-iodo anthranilic acid,acmc-1ays8,2-amino-5-iodobenzoicacid,5-iodo-2-aminobenzoic acid
72911
2-amino-5-iodobenzoic acid
C1=CC(=C(C=C1I)C(=O)O)N
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 219.0°C to 221.0°C
Color Beige or Gray to Purple
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula H2NCH3(I)CO2H
Beilstein 14, 373
Formula Weight 263.03
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Amino-5-iodobenzoic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-205-7