Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-iodopyridine 98.0+%, TCI America™
SDP

Catalog No. A184225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A184225G 25 g
A18421G 1 g
A18425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A184225G Supplier TCI America Supplier No. A184225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20511-12-0
Molecular Formula C5H5IN2
Molecular Weight (g/mol) 220.013
InChI Key IVILGUFRMDBUEQ-UHFFFAOYSA-N
Synonym 2-amino-5-iodopyridine, 2-pyridinamine, 5-iodo, 5-iodo-pyridin-2-ylamine, 5-iodo-2-pyridinamine, 5-iodo-2-aminopyridine, 2-amino-5-iodo pyridine, pyridine, 2-amino-5-iodo, 5-iod-2-pyridinamin, 5-iodo-2-pyridylamine, 6-amino-3-iodopyridine
PubChem CID 88579
IUPAC Name 5-iodopyridin-2-amine
SMILES C1=CC(=NC=C1I)N

Specifications

Melting Point 127°C
Quantity 25 g
Formula Weight 220.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-iodopyridine
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.