Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-nitro-4-picoline, 98%, Thermo Scientific™

Catalog No. ac319560010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 21901-40-6
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00010692
InChI Key GRBBNZYMXKTQAI-UHFFFAOYSA-N
Synonym 2-amino-4-methyl-5-nitropyridine, 2-amino-5-nitro-4-picoline, 2-amino-4-methyl-5-nitro pyridine, 4-methyl-5-nitro-2-pyridinamine, 4-methyl-5-nitro-2-pyridylamine, 2-pyridinamine, 4-methyl-5-nitro, 4-methyl-5-nitro-pyridin-2-ylamine, 2-amino-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2-amin, 2-amino-5-nitro-4-picoline 2-amino-4-methyl-5-nitropyridine
PubChem CID 243165
IUPAC Name 4-methyl-5-nitropyridin-2-amine
SMILES CC1=CC(=NC=C1[N+](=O)[O-])N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 220.0°C to 225.0°C
Color Brown-Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 153.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-5-nitro-4-picoline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.