Learn More
2-Amino-5-nitrobenzophenone, 98%
Description
It is used in the determination of nitrazepam in serum by gas-liquid chromatography; application in bioavailability studies and in second-harmonic generation in aromatic organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used in the determination of nitrazepam in serum by gas-liquid chromatography; application in bioavailability studies and in second-harmonic generation in aromatic organic compounds.
Solubility
Soluble in chloroform, dichloromethane, dimethyl sulfoxide, ethyl acetate and methanol. Soluble in water 3 mg/L (20°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 166°C to 168°C |
| Quantity | 100 g |
| Solubility Information | Soluble in chloroform,dichloromethane,dimethyl sulfoxide,ethyl acetate and methanol. Soluble in water 3mg/L (20°C). |
| Formula Weight | 242.23 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Amino-5-nitrobenzophenone |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 217-207-9 |
| RTECSNumber | PC4935000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.