Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-nitropyridine, 99%

Catalog No. ac104230050
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
Packaging:
Glass bottle

CAS: 4214-76-0 | C5H5N3O2 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4214-76-0
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006325
InChI Key UGSBCCAHDVCHGI-UHFFFAOYSA-N
Synonym 2-amino-5-nitropyridine, 2-pyridinamine, 5-nitro, 5-nitro-2-aminopyridine, 5-nitro-2-pyridinamine, 2-amino-5-nitro pyridine, pyridine, 2-amino-5-nitro, 5-nitro-pyridin-2-ylamine, 5-nitro-pyridin-2-yl amine, 5-nitro-2-pyridylamine, 5-nitropyridin-2-amin
PubChem CID 77888
IUPAC Name 5-nitropyridin-2-amine
SMILES NC1=CC=C(C=N1)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 186.0°C to 190.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula NH2(C5H3N)NO2
Quantity 5 g
Beilstein 22,I,631
Solubility Information Solubility in water: 1.6g/L water (13°C).
Formula Weight 139.11
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-nitropyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 224-145-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.