Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-nitrothiazole, 97%

Catalog No. p-3756815
Click to view available options
Quantity:
25 g
100 g

CAS: 121-66-4 | C3H3N3O2S | 145.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-66-4
Molecular Formula C3H3N3O2S
Molecular Weight (g/mol) 145.14
MDL Number MFCD00005326
InChI Key MIHADVKEHAFNPG-UHFFFAOYSA-N
Synonym 2-amino-5-nitrothiazole, 5-nitrothiazol-2-amine, enheptin, entramin, 2-thiazolamine, 5-nitro, nitramin ido, amnizol soluble, enheptin-t, 5-nitro-2-thiazolamine, enheptin premix
PubChem CID 8486
ChEBI CHEBI:82386
IUPAC Name 5-nitro-1,3-thiazol-2-amine
SMILES NC1=NC=C(S1)[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 196°C to 204°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Quantity 25 g
Merck Index 15, 453
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in dilute mineral acids,1g/100g alcohol,1g/250g ether,practically insoluble in chloroform
Formula Weight 145.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Amino-5-nitrothiazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-490-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.