Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-trifluoromethyl-1,3,4-thiadiazole 98.0+%, TCI America™
SDP

Catalog No. A246225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A246225G 25 g
A24625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A246225G Supplier TCI America Supplier No. A246225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10444-89-0
Molecular Formula C3H2F3N3S
Molecular Weight (g/mol) 169.125
MDL Number MFCD00003109
InChI Key LTEUXHSAYOSFGQ-UHFFFAOYSA-N
Synonym 5-trifluoromethyl-1,3,4-thiadiazol-2-amine, 2-amino-5-trifluoromethyl-1,3,4-thiadiazole, 1,3,4-thiadiazol-2-amine, 5-trifluoromethyl, 5-trifluoromethyl-1,3,4 thiadiazol-2-ylamine, 1,3,4-thiadiazole, 2-amino-5-trifluoromethyl, 5-trifluoromethyl-1,3,4-thiadiazole-2-ylamine, zlchem 1059, pubchem14503, maybridge1_003448
PubChem CID 82634
IUPAC Name 5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
SMILES C1(=NN=C(S1)N)C(F)(F)F

Specifications

Melting Point 223°C
Color White-Yellow
Quantity 25 g
Formula Weight 169.13
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.