Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-fluorophenol, 97%

Catalog No. AAH66626MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66626MD 250 mg
AAH6662603 1 g
AAH6662606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66626MD Supplier Thermo Scientific Chemicals Supplier No. H66626MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

CAS: 53981-25-2 | C6H6FNO | 127.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53981-25-2
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD08532465
InChI Key MDIAVSZFIQWYST-UHFFFAOYSA-N
Synonym 6-fluoro-2-aminophenol, 3-fluoro-2-hydroxyaniline, 2-amino-6-fluoro-phenol, phenol, 2-amino-6-fluoro, pubchem10659, acmc-209lbm, 2-amino-6-flourophenol, phenol,2-amino-6-fluoro
PubChem CID 643562
IUPAC Name 2-amino-6-fluorophenol
SMILES C1=CC(=C(C(=C1)F)O)N

Specifications

Quantity 250 mg
UN Number UN2811
Beilstein 3603077
Formula Weight 127.12
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-6-fluorophenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.