Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-picoline, 98%

Catalog No. AC104400025
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104400025 2.5 kg Glass bottle
AC104400050 5 g Glass bottle
AC104401000 100 g Glass bottle
AC104405000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC104400025 Supplier Thermo Scientific Chemicals Supplier No. 104400025

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

CAS: 1824-81-3 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1824-81-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006331
InChI Key QUXLCYFNVNNRBE-UHFFFAOYSA-N
Synonym 2-amino-6-methylpyridine, 2-amino-6-picoline, 6-amino-2-picoline, 2-pyridinamine, 6-methyl, 6-methyl-2-pyridinamine, 6-methyl-2-pyridylamine, 6-methyl-2-aminopyridine, 2-picoline, 6-amino, 6-amino-2-methylpyridine, pyridine, 2-amino-6-methyl
PubChem CID 15765
IUPAC Name 6-methylpyridin-2-amine
SMILES CC1=CC=CC(N)=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 40°C to 45°C
Color Yellow
Boiling Point 208°C to 209°C
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 2.5 kg
Beilstein 22, V,9, 210
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohols and chlorinated solvents,sparingly soluble in toluene
Formula Weight 108.14
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2-Amino-6-picoline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
EINECSNumber 217-360-1
RTECSNumber US1885000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.