Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-9,9-diphenylfluorene 98.0+%, TCI America™
SDP

Catalog No. A27791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27791G 1 g
A27795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27791G Supplier TCI America Supplier No. A27791G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1268519-74-9
Molecular Formula C25H19N
Molecular Weight (g/mol) 333.434
MDL Number MFCD22376664
InChI Key MQRGCMXCVJPWHI-UHFFFAOYSA-N
PubChem CID 70700696
IUPAC Name 9,9-diphenylfluoren-2-amine
SMILES C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=C2C=C(C=C4)N)C5=CC=CC=C5

Specifications

Color Yellow
Quantity 1 g
Formula Weight 333.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-9,9-diphenylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.