Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzamide 98.0+%, TCI America™
SDP

Catalog No. A0262100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0262100G 100 g
A026225G 25 g
A0262500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0262100G Supplier TCI America Supplier No. A0262100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88-68-6
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00007981
InChI Key PXBFMLJZNCDSMP-UHFFFAOYSA-N
Synonym anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van
PubChem CID 6942
IUPAC Name 2-aminobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)N

Specifications

Melting Point 111°C
Color White
Quantity 100 g
Formula Weight 136.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzamide
EINECSNumber (3)-4561
RTECSNumber CU8993000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.