Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzenesulfonic Acid 99.0+%, TCI America™
SDP

Catalog No. A0266500G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g
500 g

Chemical Identifiers

CAS 88-21-1
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.186
MDL Number MFCD00007705
InChI Key ZMCHBSMFKQYNKA-UHFFFAOYSA-N
Synonym orthanilic acid, aniline-2-sulfonic acid, o-aminobenzenesulfonic acid, o-sulfanilic acid, aniline-o-sulfonic acid, 2-sulfanilic acid, aniline-o-sulphonic acid, 1-aminobenzene-2-sulfonic acid, 2-aminobenzenesulphonic acid, benzenesulfonic acid, 2-amino
PubChem CID 6926
ChEBI CHEBI:1015
IUPAC Name 2-aminobenzenesulfonic acid
SMILES C1=CC=C(C(=C1)N)S(=O)(=O)O

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 173.19
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzenesulfonic Acid
EINECSNumber (3)-1971
RTECSNumber DB4727700
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.