Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzhydrazide, 98+%

Catalog No. p-7081010
Click to view available options
Quantity:
10 g
50 g

CAS: 1904-58-1 | C7H9N3O | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1904-58-1
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.17
MDL Number MFCD00017056
InChI Key GRWMSCBKWMQPON-UHFFFAOYSA-N
Synonym 2-aminobenzhydrazide, o-aminobenzhydrazide, anthranilic acid hydrazide, benzoic acid, 2-amino-, hydrazide, anthranilic hydrazide, anthraniloyl hydrazide, anthraniloyl hydrazine, 2-aminobenzoylhydrazine, 2-aminobenzoic acid hydrazide, o-aminobenzoic hydrazide
PubChem CID 94728
IUPAC Name 2-aminobenzohydrazide
SMILES NNC(=O)C1=CC=CC=C1N

Specifications

Melting Point 122°C to 124°C
Quantity 10 g
Beilstein 742811
Formula Weight 151.17
Percent Purity ≥98%
Chemical Name or Material 2-Aminobenzhydrazide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 217-604-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.